Find methods for your needs
OSHA Method 60 for volatile amines using reversed-phase chromatography with the Thermo Scientific Acclaim C30 HPLC column
Description
The Thermo Scientific UltiMate 3000 system is applied for the analysis of volatile amines. In this work, 1-naphthyl isothiocyanate (NITC) reacts with amines in an air sampling tube, to produce a UV-absorbing thiourea derivative. In this work, the Thermo Scientific Acclaim C30 separates the three amines in OSHA Method 60 with a simple isocratic reversed-phase elution. OSHA Method 60 describes the use of normal-phase chromatography, but as shown here reversed-phase chromatography can provide equivalent or better results.| Market: | Environmental |
| Keywords: | Acclaim C30, Diethylenetriamine, Ethylenediamine, HPLC, NITC derivatives, OSHA Method 60, Triethylenetetramine, UV detection, Volatile amines |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.