Find methods for your needs
Rapid analysis of CARB-1004 DNPH aldehyde and ketone standards using a Thermo Scientific Acclaim Carbonyl C18 RSLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of CARB-1004 DNPH aldehyde and ketone standards. The separation is performed on a Thermo Scientific Acclaim Carbonyl C18 RSLC column, with UV detection at 360 nm.| Market: | Environmental |
| Keywords: | HPLC, UV detection, CARB-1004, Acclaim Carbonyl C18, Carbonyl DNPH |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.