Find methods for your needs
Fast analysis of metaraminol bitartrate using a Thermo Scientific Acclaim Trinity P1 HPLC column
Description
The Thermo Scientific UltiMate 3000 HPLC system is applied for the analysis of metaraminol bitartrate. The separation is performed on a Thermo Scientific Acclaim Trinity P1 HPLC column, with UV detection at 210 nm.| Market: | Pharma |
| Keywords: | Acclaim Trinity P1, HPLC, Metaraminol bitartrate, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.