Find methods for your needs
Rapid LC-MS/MS analysis of Acetylcholinesterase inhibitors in plasma using Thermo Scientific SOLA WCX SPE cartridges and a Thermo Scientific Syncronis HILIC HPLC column
Description
The Thermo Scientific Dionex UltiMate 3000 RSLC system is applied for the analysis of edrophonium, neostigmine, and pyridostigmine in plasma. The samples were prepared using Thermo Scientific SOLA WCX solid phase extraction (SPE) cartridges and the separation performed on a Thermo Scientific Syncronis HILIC HPLC column, with MS/MS detection. For Research Use Only. Not for use in diagnostic procedures.| Market: | Pharma |
| Keywords: | Acetylcholinesterase inhibitors, Benzyldimethylphenylammonium chloride, Edrophonium, LC-MS/MS, MS detection, Neostigmine, Pyridostigmine, SOLA WCX, SPE, Syncronis HILIC |
| Matrix: | Plasma |
For Research Use Only. Not for use in diagnostic procedures.