Find methods for your needs
Improved analysis of cefepime and related substances using a Thermo Scientific Acclaim 120 C18 HPLC column
Description
The Thermo Scientific UltiMate 3000 HPLC system is applied for the determination of cefepime and related substances. The separation is performed on a Thermo Scientific Acclaim 120 C18 HPLC column, with UV detection at 254 nm.| Market: | Pharma |
| Keywords: | Acclaim 120 C18, Cefepime, HPLC, Impurities, UltiMate 3000, UV detection |
| Matrix: | Powder |
For Research Use Only. Not for use in diagnostic procedures.