Find methods for your needs
Improved analysis of melamine and metformin hydrochloride using a Thermo Scientific BioBasic SCX HPLC column
Description
The Thermo Scientific HPLC system is applied for the determination of melamine and metformin hydrochloride. The separation is performed on a Thermo Scientific BioBasic SCX HPLC column, with UV detection at 218 nm.| Market: | Pharma |
| Keywords: | BioBasic SCX, HPLC, melamine, Metformin, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.