Find methods for your needs
Rapid LC-MS/MS analysis of three biopterins on a Thermo Scientific Syncronic HILIC HPLC column
Description
A liquid chromatography method for the analysis of Biopterin, Dihydro-biopterin and Tetrahydro-biopterin was developed on a Thermo Scientific Syncronis HILIC HPLC column for a fast separation with a cycle time of 3 minutes while maintaining excellent peak shape and resolution.| Market: | Pharma |
| Keywords: | HPLC, MS detection, Syncronis HILIC, Biopterins |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.