Find methods for your needs
Improved analysis of diquat and paraquat herbicides using a Thermo Scientific Acclaim Trinity Q1 (2.1 mm ID) HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of diquat and paraquat herbicides. The separation is performed on a Thermo Scientific Acclaim Trinity Q1 (2.1 mm ID) HPLC column, with UV detection at 290 nm.| Market: | Environmental |
| Keywords: | Acclaim Trinity Q1, Diquat, Herbicides, HPLC, Paraquat, UV detection |
| Matrix: | Drinking water |
For Research Use Only. Not for use in diagnostic procedures.