Find methods for your needs
Fast analysis of amlodipine in human plasma using Thermo Scientific SOLA CX SPE and a Thermo Scientific Accucore RP-MS HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of amlodipine in human plasma. The samples are prepared using a Thermo Scientific SOLA CX solid phase extraction (SPE) plate, and the separation is performed on a Thermo Scientific Accucore RP-MS HPLC column, with MS detection. For Research Use Only. Not for use in diagnostic procedures.Market: | Pharma |
Keywords: | Accucore RP-MS, Amlodipine, LC-MS/MS, MS detection, Nifedipine, SOLA CX, SPE |
Matrix: | Human plasma |
For Research Use Only. Not for use in diagnostic procedures.