Find methods for your needs
Rapid analysis of explosives using a Thermo Scientific Accucore C18 HPLC column
Description
The Thermo Scientific HPLC system is applied for the determination of explosives. The separation is performed on a Thermo Scientific Accucore C18 HPLC column, with UV detection at 254 nm. The separation achieved on the Thermo Scientific Accucore C18 HPLC column is compared to the separation achieved on a Thermo Scientific Accucore PFP HPLC column.| Market: | Chemical |
| Keywords: | Accucore C18, Comparison, Dinitrobenzene, Dinitrotoluene, Explosives, HPLC, Nitrobenzene, Trinitrobenzene, Trinitrotoluene, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.