Find methods for your needs
Fast analysis of phenols using conventional GC instrumentation Be the first to rate this application
fast analysis phenols using conventional gc instrumentation
Description
In this work the performance of a 0.15 mm ID column is compared to a 0.25 mm ID column, and an increase in speed of analysis for a phenol standard mix is demonstrated according to EPA Method 625. There was no compromise in the separation capability of the method and conversion of the conventional method was easily achieved.
Market: Environmental
Keywords: EPA Method 625, Flame ionisation detection (FID), GC, Phenols, TraceGOLD TG-5MS
Matrix: N/A
Uploaded on 9/4/2012.
For Research Use Only. Not for use in diagnostic procedures.