Description
The Thermo Scientific Dionex UltiMate 3000 HPLC system is applied for the determination of decitabine and its α-anomer. The separation is performed on a Thermo Scientific Accucore HILIC HPLC column which provides enhanced retention of polar and hydrophilic analytes.| Market: | Pharma |
| Keywords: | Accucore HILIC, Decitabine, HILIC, HPLC, Impurities, UV detection |
| Matrix: | N/A |
| Author: | Prakash Chander |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.