Find methods for your needs
GC-MS analysis of diethylene glycol and ethylene glycol in toothpaste using a Thermo Scientific TraceGOLD TG-WaxMS column
Description
The Thermo Scientific GC system is applied for the analysis of diethylene glycol and ethylene glycol in toothpaste. The separation is performed using a Thermo Scientific TraceGOLD TG-WaxMS column with 5m SafeGuard.| Market: | Other |
| Keywords: | Diethylene glycol, Ethylene glycol, GC/MS, MS detection, SafeGuard, TraceGOLD TG-WaxMS |
| Matrix: | Toothpaste |
For Research Use Only. Not for use in diagnostic procedures.