Find methods for your needs
Improved analysis of cyclic monophosphates using a Thermo Scientific Hypercarb HPLC column
Description
The Thermo Scientific HPLC system is applied for the determination of cyclic monophosphates. The separation is performed on a Thermo Scientific Hypercarb HPLC column, with UV detection at 254 nm.| Market: | Chemical | 
| Keywords: | Adenosine, Cyclic monophosphates, Cytidine, Guanosine, HPLC, Hypercarb, Nucleotides, Uridine, UV detection | 
| Matrix: | N/A | 
For Research Use Only. Not for use in diagnostic procedures.