Find methods for your needs
Improved analysis of PTH amino acids using a Thermo Scientific Hypersil GOLD HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of PTH amino acids. The separation is performed on a Thermo Scientific Hypersil GOLD HPLC column, with UV detection at 269 nm.| Market: | Food and Beverage |
| Keywords: | Analine, Asparagine, Aspartic acid, Glutamic acid, HPLC, Hypersil GOLD, Leucine, Methionine, Phenylanaline, PTH amino acids, Serine, Tryptophan, Tyrosine, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.