Find methods for your needs
Fast analysis of anaesthetics using a Thermo Scientific Hypersil GOLD HPLC column Be the first to rate this application
fast analysis anaesthetics using a thermo scientific hypersil gold hplc column
Description
The Thermo Scientific HPLC system is applied for the analysis of anaesthetics. The separation is performed on a Thermo Scientific Hypersil GOLD HPLC column, with UV detection at 220 nm.
Market: Pharma
Keywords: Anaesthetics, Benzocaine, HPLC, Hypersil GOLD, Lidocaine, Tetracaine, UV detection
Matrix: N/A
Uploaded on 9/3/2012.
For Research Use Only. Not for use in diagnostic procedures.