Find methods for your needs
Improved analysis of catechins using a Thermo Scientific Accucore XL C8 4 µm HPLC column
Description
The Thermo Scientific HPLC system is applied for the determination of catechins. The separation is performed on a Thermo Scientific Accucore C18 XL HPLC column, with UV detection at 280 nm.| Market: | Food and Beverage |
| Keywords: | Accucore XL C8, Catechin, Catechin gallate, Epicatechin gallate, Epigallocatechin, Epigallocatechin gallate, Gallocatechin gallate, HPLC, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.