Find methods for your needs
Improved analysis of 5-monophosphates using a Thermo Scientific Hypersil GOLD aQ HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of 5-monophosphates. The separation is performed on a Thermo Scientific Hypersil GOLD aQ HPLC column, with UV detection at 254 nm.| Market: | Chemical |
| Keywords: | 5-AMP, 5-CMP, 5-GMP, 5-monophosphates, 5-UMP, HPLC, Hypersil GOLD aQ, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.