Find methods for your needs
Fast analysis of testosterones using a Thermo Scientific Hypersil GOLD HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of testosterones. The separation is performed on a Thermo Scientific Hypersil GOLD HPLC column, with UV detection at 254 nm.| Market: | Pharma |
| Keywords: | 11-ketotestosterone, 19-Nortestosterone, Epitestosterone, HPLC, Hypersil GOLD, Nandralone, Testosterones, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.