Find methods for your needs
Fast analysis of salmeterol xinafoate using a Thermo Scientific Hypersil GOLD HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of salmeterol xinafoate. The separation is performed on a Thermo Scientific Hypersil GOLD HPLC column, with UV detection at 254 nm.| Market: | Pharma | 
| Keywords: | Asthma, Bronchodilator, HPLC, Hypersil GOLD, Salmeterol xinafoate, UV detection | 
| Matrix: | N/A | 
For Research Use Only. Not for use in diagnostic procedures.