Find methods for your needs
Fast analysis of arbutin using a Thermo Scientific Hypersil GOLD aQ HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of arbutin. The separation is performed on a Thermo Scientific Hypersil GOLD aQ HPLC column, with UV detection at 205 nm.Market: | Pharma |
Keywords: | Arbutin, HPLC, Hypersil GOLD aQ, UV detection |
Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.