Find methods for your needs
Improved analysis of glycosides using a Thermo Scientific Hypersil GOLD aQ HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of glycosides. The separation is performed on a Thermo Scientific Hypersil GOLD aQ HPLC column, with UV detection at 250 nm.Market: | Chemical |
Keywords: | Baicalein, Barbaloin, Digitoxin, Esculin, Glycosides, Glycyrrhizic acid, HPLC, Hypersil GOLD aQ, Ouabain, Salicin, UV detection |
Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.