Find methods for your needs
Fast analysis of uron herbicides using a Thermo Scientific Hypersil GOLD HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of uron herbicides. The separation is performed on a Thermo Scientific Hypersil GOLD 1.9 µm HPLC column, with UV detection at 254 nm.| Market: | Environmental |
| Keywords: | Chlortoluron, Diuron, HPLC, Hypersil GOLD, Metoxuron, Monuron, Tebuthiuron, Uron herbicides, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.