Find methods for your needs
Improved analysis of itraconazole using a Thermo Scientific Hypersil GOLD HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of itraconazole. The separation is performed on a Thermo Scientific Hypersil GOLD HPLC column, with UV detection at 254 nm.| Market: | Pharma |
| Keywords: | antifungal, HPLC, Hypersil GOLD, Itraconazole, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.