Find methods for your needs
Fast analysis of anti-psychotics using a Thermo Scientific Hypersil GOLD HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of anti-psychotics. The separation is performed on a Thermo Scientific Hypersil GOLD HPLC column, with UV detection at 254 nm.| Market: | Pharma |
| Keywords: | Anti-psychotics, Chlorpromazine, HPLC, Hypersil GOLD, Promazine, Propionylpromazine, Triflupromazine, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.