Find methods for your needs
Robust analysis of polycyclic aromatic hydrocarbons (PAHs) using a Thermo Scientific TraceGOLD TG-5MT metal GC column
Description
In this work the Thermo Scientific TraceGOLD TG-5MT metal GC column is used for the analysis of PAHs. The selectivity of the TG-5MT column was found to be very similar to the TraceGOLD TG-5MS column. The advantage of using the metal column is the higher maximum operating temperature which means a high temperature 'bake-out' can be performed at the end of the analysis to remove any highly retained contaminants, ensuring a clean system is maintained.| Market: | Environmental |
| Keywords: | GC, MS getection, Polycyclic aromatic hydrocarbons (PAHs), TraceGOLD TG-5MT |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.