Find methods for your needs
Fast analysis of mepiquat using a Thermo Scientific BioBasic SCX HPLC column Be the first to rate this application
fast analysis mepiquat using a thermo scientific biobasic scx hplc column
Description
The Thermo Scientific HPLC system is applied for the determination of mepiquat. The separation is performed on a Thermo Scientific BioBasic SCX HPLC column, with MS detection.
Market: Environmental
Keywords: BioBasic SCX, HPLC, Mepiquat, MS detection
Matrix: N/A
Uploaded on 8/31/2012.
For Research Use Only. Not for use in diagnostic procedures.