Find methods for your needs
Improved analysis of chlorophenols using a Thermo Scientific Hypersil GOLD PFP HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of chlorophenols. The separation is performed on a Thermo Scientific Hypersil GOLD PFP HPLC column, with UV detection at 230 nm| Market: | Chemical |
| Keywords: | Chlorophenol, HPLC, Hypersil Gold PFP, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.