Find methods for your needs
Fast analysis of creatine and creatinine on a Thermo Scientific Hypercarb HPLC column Be the first to rate this application
Description
The Thermo Scientific HPLC system is applied for the determination of creatine and creatinine. The separation is performed on a Thermo Scientific Hypercarb HPLC column, with UV detection at 216 nm.
Market: Other
Keywords: Creatine, Creatinine, HPLC, Hypercarb, UV detection
Matrix: N/A
Uploaded on 8/30/2012.
For Research Use Only. Not for use in diagnostic procedures.