Find methods for your needs
Rapid analysis of artesunate and dihydroartemisinin using a Thermo Scientific Accucore RP-MS HPLC column Be the first to rate this application
Description
The Thermo Scientific HPLC system is applied for the determination of artesunate and dihydroartemisinin. The separation is performed on a Thermo Scientific Accucore RP-MS HPLC column, with UV detection at 210 nm.
Market: Pharma
Keywords: Accucore RP-MS, Artesunate, Dihydroartemisinin, HPLC, UV detection
Matrix: N/A
Uploaded on 8/30/2012.
For Research Use Only. Not for use in diagnostic procedures.