Find methods for your needs
Improved analysis of polybrominated diphenyl ethers (PBDEs) using a Thermo Scientific Hypersil GOLD Phenyl UHPLC column
Description
The Thermo Scientific HPLC system is applied for the determination of polybrominated diphenyl ethers (PBDEs). The separation is performed on a Thermo Scientific Hypersil GOLD Phenyl UHPLC column, with UV detection at 230 nm.| Market: | Environmental |
| Keywords: | HPLC, Hypersil GOLD Phenyl, PBDE, Polybrominated diphenyl ethers, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.