Find methods for your needs
Fast analysis of contaminants in soil using a Thermo Scientific Hypersil GOLD C8 HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of contaminants in soil. The separation is performed on a Thermo Scientific Hypersil GOLD C8 HPLC column, with UV detection at 220 nm.| Market: | Environmental |
| Keywords: | Benzene, Benzoic acid, Benzyl alcohol, Contaminants, HPLC, Hypersil GOLD C8, Nitrobenzene, p-Hydroxybenzaldehyde, soil, UV detection |
| Matrix: | Soil |
For Research Use Only. Not for use in diagnostic procedures.