Find methods for your needs
Fast analysis of β-Carotene using a Thermo Scientific Hypersil GOLD C8 HPLC column Be the first to rate this application
Description
The Thermo Scientific HPLC system is applied for the analysis of β-Carotene. The separation is performed on a Thermo Scientific Hypersil GOLD C8 HPLC column, with UV detection at 450 nm.
Market: Food and Beverage
Keywords: B-Carotene, carotenoid, HPLC, Hypersil GOLD C8, Lutein, Lycopene, UV detection
Matrix: N/A
Uploaded on 8/29/2012.
For Research Use Only. Not for use in diagnostic procedures.