Find methods for your needs
Fast analysis of estrogens using a Thermo Scientific Hypersil GOLD HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of estrogens. The separation is performed on a Thermo Scientific Hypersil GOLD HPLC column in less than 5 minutes, with UV detection at 280 nm.| Market: | Pharma |
| Keywords: | Estradiol, Estriol, Estrogens, Estrone, HPLC, Hypersil GOLD, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.