Find methods for your needs
Rapid analysis of analgesic tablets using a Thermo Scientific Hypersil GOLD HPLC column Be the first to rate this application
Description
The Thermo Scientific HPLC system is applied for the analysis of analgesic tablets. The separation is performed on a Thermo Scientific Hypersil GOLD HPLC column in less than 5 minutes, with UV detection at 230 nm.
Market: Pharma
Keywords: Analgesics, Caffeine, Codeine, HPLC, Hypersil GOLD, Paracetamol, Solpadeine, Tablet, UV detection
Matrix: Tablet
Uploaded on 8/28/2012.
For Research Use Only. Not for use in diagnostic procedures.