Find methods for your needs
Rapid determination of Melamine in powdered milk by LC-MS/MS using Thermo Scientific HyperSep Retain CX SPE and a Thermo Scientific Accucore HILIC HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of Melamine in powdered milk. The sample was prepared using HyperSep Retain CX solid phase extraction (SPE) cartridges, and the separation performed on a Thermo Scientific Accucore HILIC HPLC column in less than 2 minutes, with MS/MS detection.| Market: | Food and Beverage |
| Keywords: | Accucore HILIC, HyperSep Retain CX, LC-MS/MS, melamine, MS detection, Powdered milk, SPE |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.