 Rate
                        Rate
                     Ask the Expert
                        Ask the Expert
                     Suggest an Application
                            Suggest an Application
                         Add to Favorites
                        Add to Favorites
                     Create
                            CreateParts List
 Tell a
                        Tell a Friend
 Print
                        PrintFind methods for your needs
Fast determination of Tenofovir from human plasma by LC-MS/MS using Thermo Scientific SOLA SCX SPE and a Thermo Scientific Hypersil GOLD HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of Tenofovir from human plasma. The sample was prepared using SOLA SCX solid phase extraction (SPE), and the separation performed on a Thermo Scientific Hypersil GOLD HPLC column in less than 3 minutes, with MS/MS detection. For Research Use Only. Not for use in diagnostic procedures.| Market: | Pharma | 
| Keywords: | Antiretroviral, Hypersil GOLD, LC-MS/MS, MS detection, Nucleotide analogue reverse transcriptase, Plasma, SOLA SCX, SPE, Tenofovir | 
| Matrix: | Human plasma | 
For Research Use Only. Not for use in diagnostic procedures.