Find methods for your needs
Demonstration of improved efficiency, sensitivity and resolution for the analysis of endocrine disruptors using a solid core C8 4 µm HPLC column
Description
The Thermo Scientific Dionex UltiMate 3000 RSLC system is applied for the analysis of endocrine disruptors. The separation is performed on a Thermo Scientific Accucore XL C8 4 µm HPLC column, with UV detection at 220 nm. Compared to a conventional 5 μm fully porous column using the same experimental conditions, the Accucore XL C8 4 μm HPLC column provides improved efficiency, sensitivity and resolution for the analysis of these compounds.| Market: | Pharma |
| Keywords: | Accucore XL C8, Atrazine, Bisphenol A, Desethyl Atrazine, Diuron, Endocrine disruptors, HPLC, Simazine, UV detection |
| Matrix: | N/A |
| Author: | Eilidh MacRitchie |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.