Find methods for your needs
Improved analysis of nineteen explosives using a Thermo Scientific Hypersil GOLD aQ HPLC column Be the first to rate this application
Description
The Thermo Scientific HPLC system is applied for the analysis of 19 explosives. The separation is performed on a Thermo Scientific Hypersil GOLD aQ HPLC column, with UV detection at 245 and 215 nm.
Market: Chemical
Keywords: Explosives, HPLC, Hypersil GOLD aQ, UV detection
Matrix: N/A
Uploaded on 8/28/2012.
For Research Use Only. Not for use in diagnostic procedures.