Find methods for your needs
Fast LC-MS/MS method for the determination of Paclitaxel in human serum using Thermo Scientific SOLA HRP SPE and a Thermo Scientific Accucore RP-MS HPLC column
Description
The Thermo Scientific UltiMate 3000 RSLC system is applied for the analysis of Paclitaxel from human serum. The sample was prepared using SOLA HRP solid phase extraction (SPE), and the separation performed on a Thermo Scientific Accucore RP-MS HPLC column in less than 3 minutes, with MS/MS detection. For Research Use Only. Not for use in diagnostic procedures.| Market: | Pharma |
| Keywords: | Accucore RP-MS, Docetaxel, LC-MS/MS, MS detection, Paclitaxel, SOLA HRP, SPE |
| Matrix: | Human serum |
For Research Use Only. Not for use in diagnostic procedures.