Find methods for your needs
Fast analysis of Choline and Acetylcholine by LC-MS/MS using a Thermo Scientific Syncronis HILIC HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of choline and acetylcholine. The separation is performed on a Thermo Scientific Syncronis HILIC 1.7 μm HPLC column in less than 2.5 minutes, with MS/MS detection.| Market: | Pharma |
| Keywords: | Acetylcholine, choline, LC-MS/MS, MS detection, Syncronis HILIC |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.