Find methods for your needs
Rapid LC-MS/MS method for rosuvastatin using Thermo Scientific SOLA SPE and a Thermo Scientific Accucore PFP HPLC column Be the first to rate this application
Description
The Thermo Scientific HPLC system is applied for the analysis of rosuvastatin and its degradation products. The sample is prepared using a Thermo Scientific SOLA SPE plate, and the separation is performed on a Thermo Scientific Accucore RP-MS HPLC column in less than 3.6 minutes, with MS/MS detection. For Research Use Only. Not for use in diagnostic procedures.
Market: Pharma
Keywords: Accucore RP-MS, Degradation Products, Human Plasma, LC-MS/MS, MS detection, Rosuvastatin, SOLA, SPE
Matrix: Human plasma
Uploaded on 8/24/2012.
For Research Use Only. Not for use in diagnostic procedures.