Find methods for your needs
Rapid analysis of zafirlukast in human plasma using Thermo Scientific HyperSep Retain AX SPE and a Thermo Scientific Accucore RP-MS HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of zafirlukast. The sample are prepared using a Thermo Scientific HyperSep Retain AX solid phase extraction (SPE) plate, and the separation is performed on a Thermo Scientific Accucore RP-MS HPLC column in less than 3 minutes, with MS/MS detection. For Research Use Only. Not for use in diagnostic procedures.| Market: | Pharma |
| Keywords: | Accucore RP-MS, Asthma, Human Plasma, HyperSep Retain AX, LC-MS/MS, MS detection, SPE, Zafirlukast |
| Matrix: | Human plasma |
For Research Use Only. Not for use in diagnostic procedures.