Find methods for your needs
Related substance analysis of Estradiol Valerate as per EP using Accucore C18 HPLC column
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied for the analysis of Related substance of Estradiol Valerate as per EP. The analysis was performed on a Thermo Scientific Accucore C18 HPLC column using the method described in the EP monograph. The results obtained were found to be within acceptance criteria as stated in the monograph. Resolution between Impurity C & estradiol valerate peak was observed to be 3.18 in the refernce solution, a (Criteria is NLT 2.5)| Market: | Pharma |
| Keywords: | RS; Estradiol Valerate; EP monograph; HPLC; UV |
| Matrix: | N/A |
| Author: | Dr. Sachin Pandey |
| Affiliation: | Thermo Fisher Scientific, APAC CoE Lab, Ahmedabad, India |
For Research Use Only. Not for use in diagnostic procedures.