Find methods for your needs
AN352: Rapid Determination of Persistent Organic Pollutants (POPs) Using Accelerated Solvent Extraction (ASE)
Description
The following compounds are listed by UNEP to be POPS: • Pesticides: Aldrin, Chlordane, DDT, Dieldrin, Endrin, Heptachlor, Mirex, and Toxaphene • Industrial chemicals: Hexachlorobenzene, and PCB (polychlorinated biphenyl) • Chemical by-products (Dioxins): Polychlorinated dibenzo-p-dioxins and polychlorinated dibenzofurans (PCDD and PCDF) This application note describes methods and results for extraction of the POPs listed above, with tables comparing ASE to traditional extraction methods.| Market: | Environmental |
| Keywords: | GC, GC/MS, Pesticides, ASE, Soils, Hexachlorobenzene, Heptachlor, Aldrin, Dieldrin, Endrin, ASE 200, ASE 350, Persistent Organic Pollutants (POPs), PCB (polychlorinated biphenyl), Dioxins (PCDD and PCDF), Gamma Chlordane, Alpha Chlordane, p,p’-DDT |
| Matrix: | Sediments, Semisolid waste, Sludges, Soils, Solid waste |
For Research Use Only. Not for use in diagnostic procedures.