Find methods for your needs
AB875: Modification of EPA Method 608 for Automated Solid Phase Extraction (SPE) of Wastewaters and Surface Waters Using the Dionex AutoTrace Instrument
Description
EPA Method 525.2, Determination of Organic Compounds in Drinking Water by Liquid-Solid Extraction and Capillary Column Gas Chromatography/Mass Spectrometry, can be used as a basis for developing a liquid-solid Extraction method for EPA Method 608, Methods for Organic Chemical Analysis of Municipal and Industrial Wastewater. Unlike the drinking water methods, however, analysis of wastewaters and the streams they flow into, need to contend with particulates. The Dionex AutoTrace 280 instrument has features that allow analysis of samples even with high amounts of suspended solids.| Market: | Environmental |
| Keywords: | ddd, Solid phase extraction, SPE, Heptachlor, Methoxychlor, Aldrin, Endosulfan I, Dieldrin, Endrin, Endosulfan II, Endosulfan sulfate, Alpha-BHC, Gamma-BHC, Beta-BHC, delta-BHC, DDT, AutoTrace 280, DDE, Hepta.epoxide, SS:DCBP, SS:TCX |
| Matrix: | Surface water, Waste water |
| Author: | David Larabee-Zierath |
| Affiliation: | University Hygienic Laboratory, Iowa's Environmental and Public Health Laboratory, Iowa City, IA |
For Research Use Only. Not for use in diagnostic procedures.