Find methods for your needs
AN347: Use of Accelerated Solvent Extraction (ASE) for Cleaning and Elution of XAD Resin
Description
XAD resins are used as solid phase extraction materials for applications in water and air sampling. When received, resins are wet and contain contaminants. The current methods for cleaning and eluting require large amounts of solvent and time. Here, an XAD-2 resin is cleaned using ASE, then spiked with organochlorine pesticides (OCP) and polyaromatic hydrocarbons (PAH) standards and eluted using ASE. The results show that ASE effectively and efficiently cleans and dries the resin without damaging the particles, and is able to elute OCP and PAH compounds from XAD resin with good results.| Market: | Material Science |
| Keywords: | ASE, ASE 200, ASE 350, XAD resins, Soxhlet extractions, organochlorine pesticides (OCP), polyaromatic hydrocarbons (PAH) |
| Matrix: | Resins |
For Research Use Only. Not for use in diagnostic procedures.