Find methods for your needs
AN349: Rapid Determination of Organochlorine Pesticides in Animal Feed Using Accelerated Solvent Extraction (ASE)
Description
ASE is EPA approved under method 3545A. This technique uses high temperatures and pressures to increase the kinetics of the extraction process, thus decreasing the extraction time and solvent consumption. Also, because ASE is automated, it allows unattended extraction of up to 24 samples. In this application note, OCPs are extracted from certified reference material (CRM) BCR 115 (Institute for Reference Materials and Measurement, Geel, Belgium), an animal feed containing certified levels of organochlorine pesticides.| Market: | Food and Beverage |
| Keywords: | Animal feed, ASE, Organochlorine pesticides, Heptachlor, Aldrin, Dieldrin, Endrin, ASE 200, ASE 350, EPA 3545A, α-HCH, HCB, β-HCH, γ-HCH, p,p’ –DDE, p,p’ –DDD, o,p’ –DDT, p,p’ –DDT |
| Matrix: | Animal feed |
For Research Use Only. Not for use in diagnostic procedures.