Find methods for your needs
AN331: Accelerated Solvent Extraction (ASE) of Additives from Polymer Materials
Description
Accelerated Solvent Extraction (ASE) is a powerful technique that can be used to reliably extract additives from polymer materials. ASE uses organic solvents at temperatures above their atmospheric pressure boiling points to deliver extractions equivalent to traditional extraction techniques, but with faster extraction times, reduced solvent use, and automation of the extraction process. ASE is recognized as an official extraction method in U.S. EPA Method 3545.| Market: | Material Science |
| Keywords: | Additives, HPLC, ASE, UV Absorbance Detection, ASE 200, ASE 350, EPA 3545, Polymer Materials |
| Matrix: | Polymer materials |
For Research Use Only. Not for use in diagnostic procedures.