 Rate
                        Rate
                     Ask the Expert
                        Ask the Expert
                     Suggest an Application
                            Suggest an Application
                         Add to Favorites
                        Add to Favorites
                     Create
                            CreateParts List
 Tell a
                        Tell a Friend
 Print
                        PrintFind methods for your needs
AN318: Extraction of Chlorinated Herbicides Using Accelerated Solvent Extraction (ASE)
                                Description
The use of ASE in the extraction of chlorinated herbicides from solid wastes is more convenient, faster, and less solvent-intensive than previous methods.U.S. In the method reported in this application note, chlorinated herbicide recoveries by ASE are equivalent to recoveries from the wrist-shaking method recommended in Method 8150A. The procedures described in this application note meet the requirements for sample extraction as prescribed by U.S. EPA Method 3545.| Market: | Environmental | 
| Keywords: | ASE, Soils, Solid Wastes, ASE 200, ASE 350, EPA 3545, chlorinated herbicides | 
| Matrix: | Soil | 
For Research Use Only. Not for use in diagnostic procedures.